n-Pentan ≥99%, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®

Leverandør: Avantor

Synonymer: Pentane

5281.2500EA 3425 NOK
5281.2500
n-Pentan ≥99%, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
n-Pentane
Formel: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Smeltepunkt: -130
Flash punkt: -40
MDL nummer: MFCD00009498
CAS-nummer: 109-66-0
FN: 1265
ADR: 3 II
Merck Indeks: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (by GC) min. 99.0 %
Acidity max. 0.0005 meq/g
Evaporation Residue max. 5 ppm
GC sensitive impurities (lindane GC-ECD) max. 5 ng/l
Water (KF) max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR