1-Heksansulfonsyre natriumsalt monohydrat concentrate for dilution to 0.005 M in HPLC grade water for ion pair chromatography, Supelco®

Leverandør: Merck

Synonymer: Sodium 1-hexanesulphonate monohydrate

52864-6X1AMP-F
52864-6X1AMP-FEA 2350 NOK
52864-6X1AMP-F
1-Heksansulfonsyre natriumsalt monohydrat concentrate for dilution to 0.005 M in HPLC grade water for ion pair chromatography, Supelco®
1-Heksansulfonsyre natriumsalt monohydrat

Dilute contents of one ampule (~0.3 M) to 1 L with HPLC grade water to obtain a 0.005 M eluent solution.

Formel: C₆H₁₃NaO₃S·1H₂O
MW: 206.24 g/mol
MDL nummer: MFCD00149549
CAS-nummer: 207300-91-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR