Sentrifuge, vakuum, Concentrator plus
Leverandør: EPPENDORF
5305000.568 5305000.100 5305000.703 5305000.762 5305000.169 5305000.363 5305000.304 5305000.509
EPPE5305000.568EA 0 NOK
EPPE5305000.568 531-0888 531-0889 EPPE5305000.762 EPPE5305000.169 EPPE5305000.363 531-0872 531-0873
Sentrifuge, vakuum, Concentrator plus
Fordampere Sentrifugal fordampere
This ergonomic, intuitive concentrator provides quick, efficient and gentle vacuum concentration of DNA/RNA, nucleotides, proteins and other liquid or wet samples. The coated lid provides superior chemical resistance against aggressive acids and organic solvents (e.g. TFA, DMSO).
  • Chemical resistant, maintenance-free PTFE diaphragm pump eliminates the need for changing pump oil
  • Very quiet operation (<50 dB(A)), small footprint
  • Simple operation due to intuitive control panel
  • Maintenance-free induction drive and chemical-resistant, stainless steel chamber