VWR® Evaporating Flasks
Leverandør: VWR Collection
531-1099EA 240 NOK
531-1099 531-1100 531-1101 531-1102 531-1103
VWR® Evaporating Flasks
Kolber Kolber, rotasjonsfordamper
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance and high resistance to thermal shock
  • White labelling area and imprinted catalogue number
These pear-shaped rotary evaporating flasks are produced from thick walled borosilicate glass tubing. With standard ground socket, suitable for all standard rotary evaporators.