Supelco® Organic Reference Standard, cis-4,7,10,13,16,19-Docosahexaenoic acid

Leverandør: Merck

53171-10MG
53171-10MGEA 865 NOK
53171-10MG
Supelco® Organic Reference Standard, cis-4,7,10,13,16,19-Docosahexaenoic acid
Standarder Organiske standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d57dbe356a03263d0a74399b7e3974cdccebdc3&vid=1d57dbe356a03263d0a74399b7e3974cdccebdc3&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) DHA
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 6217-54-5
MDL No. MFCD00065722
Molekylvekt 328,49
Flammepunkt 62 °C - Closed cup
Fraktforhold DICE
Lagringsforhold –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR