Imhofftrakt, Sedimentasjonstrakt, Nalgene®
Leverandør: Thermo Fisher Scientific
1000-0010
532-0101EA 1075 NOK
532-0101
Imhofftrakt, Sedimentasjonstrakt, Nalgene®
Trakter Immhoff-trakter
PC with PP autoclavable stopper.
  • As transparent as glass
  • Easy draining via screw plug
  • Leakproof closure, closure design allows user to read graduations to zero
Graduation: 0 to 1 ml/0,1; 1 to 10 ml/0,5; 10 to 20 ml/1; 20 to 40 ml/2; 40 to 100 ml/10; 100 to 500/50; 500 to 1000 ml/100.