Organic reference standard, Propylamine, analytical standard, Supelco®

Leverandør: Merck

53646-1ML
53646-1MLEA 560 NOK
53646-1ML
Organic reference standard, Propylamine, analytical standard, Supelco®
Standarder Organiske standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d1979adc611c96b12bdcf0df9f2b679ef169bcd&vid=9d1979adc611c96b12bdcf0df9f2b679ef169bcd&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Beskrivelse Propylamine
Environmentally Preferable
CAS-nr 107-10-8
Formel C₃H₉N
Smeltepunkt –83 °C
kokepunkt 48 °C
Flammepunkt –30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR