Soxhlet extractors, complete assemblies
Leverandør: SciLabware
100RASX 200RASX 600RDSX
537-0022EA 5275 NOK
537-0022 537-0023 BIBB600RDSX
Soxhlet extractors, complete assemblies
Ekstraktorer Soxhlet- og Soxhlet-typeuttrekkere
PYREX®, borosilicate glass.
  • Complete glass assembly
  • Low solvent volumes required for extraction of soluble components in solids
  • Range of sizes of extractor to meet different sample sizes
* 75 mm diameter extractor flange.
Leveringsinformasjon: Assembly consists of round bottom flask, Soxhlet extractor and Allihn condenser.