1,4-Dioksan, AR®, Laboratory Reagent, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials

5413-25EA 1600 NOK
5413-25
1,4-Dioksan, AR®, Laboratory Reagent, Macron Fine Chemicals™
1,4-Dioksan
Formel: C₄H₈O₂
MDL nummer: MFCD00006571
CAS-nummer: 123-91-1
FN: 1165
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR