n-Heksan 95%, AR®, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger

5414-25EA 1475 NOK
5414-25
n-Heksan 95%, AR®, Macron Fine Chemicals™
n-Heksan
Formel: H₃C(CH₂)₄CH₃
MDL nummer: MFCD00009520
CAS-nummer: 110-54-3
FN: 1208
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Assay (GC) ≥95%
Water (KF) ≤0,02%
Titrable Acid (meq/g) ≤0,0005
Residue after Evaporation ≤0,001%
Sulfur Compounds ≤0,001%
For Laboratory, Research or Manufacturing Use


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR