Butanon, AR®, Laboratory Reagent, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials

5447-25EA 1475 NOK
5447-25
Butanon, AR®, Laboratory Reagent, Macron Fine Chemicals™
Methyl ethyl ketone
Formel: C₂H₅COCH₃
MDL nummer: MFCD00011648
CAS-nummer: 78-93-3
FN: 1193
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR