Multi-element standard solution 5 for ICP, TraceCERT®, Supelco®
Leverandør: Merck
54704-100ML
54704-100MLEA 1825 NOK
54704-100ML
Multi-element standard solution 5 for ICP, TraceCERT®, Supelco®
Standarder Kationstandarder Multi-Element Standards
27 components:
Ag, 10 mg/l; Al, 10 mg/l; Ba, 10 mg/l; Be, 10 mg/l; Bi, 10 mg/l
Ca, 100 mg/l; Cd, 10 mg/l; Co, 10 mg/l; Cr, 10 mg/l; Cs, 10 mg/l; Cu, 10 mg/L
Fe, 100 mg/l; Ga, 10 mg/l; In, 10 mg/l; K, 100 mg/l; Li, 10 mg/l
Mg, 10 mg/l; Mn, 10 mg/l; Mo, 10 mg/l; Na, 100 mg/l; Ni, 10 mg/l
Pb, 10 mg/l; Rb, 10 mg/l; Sr, 10 mg/l; Tl, 10 mg/l; V, 10 mg/l; Zn, 10 mg/l
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
M-Clarity quality level MQ300