VWR®, Other Caps for Crimp Neck ND20 with Septum
Leverandør: VWR Collection
548-0153EA 4100 NOK
548-0153
VWR®, Other Caps for Crimp Neck ND20 with Septum
Lokk Lokk til vials
PE, transparent. Combinations of PE caps with central hole 4,3 mm and septa.
For vials with bevelled neck, cap Ø×H: 22×8,4 mm.
For vials with flat neck, cap Ø×H: 22×9,1 mm.
* 6 mm central hole.