Scintillation vials, WHEATON®
Leverandør: DWK Life Sciences
227180
548-0704EA 4150 NOK
548-0704
Scintillation vials, WHEATON®
Vialer
These clear scintillation vials with screw cap are made from soda-lime glass.
  • Low background count
  • Fitted with foil lined urea cap