VWR®, Crimp Neck Vials ND13
Leverandør: VWR Collection
548-0859AEA 6975 NOK
548-0859A
VWR®, Crimp Neck Vials ND13
Vialer
Crimp Neck ND13 vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of aluminium seal styles and septa materials.
  • High performance caps/septa
  • Crimp top vials provide an excellent seal
  • Larger opening improves sample accessibility
ND13 vials with a crimp neck are available in clear version. For use in a broad range of instruments as GC and HPLC standard vials with a flat bottom. Broad variety of crimp caps mostly available with assembled stopper or septa are engineered to preventMore Product Information