VWR® WebSeal™, Kromatografi brønnplater, 96-brønns, PP, sertfifsert
Leverandør: VWR Collection
548-3371EA 925 NOK
548-3371 548-2401
VWR® WebSeal™, Kromatografi brønnplater, 96-brønns, PP, sertfifsert
Mikroplater HTS-mikroplater
PP, non coated, non sterile, chromatography tested and certified. Manufactured from ultra low bleed high purity basis resin and fully lot tested by GC/MS for organic extractables. Designed for sensitive applications, lower sample concentrations, regulated laboratories, and/or TIC and full chromatograms.
  • Microplates are chemically and thermally resistant and will tolerate temperatures from −80 to +121 °C
  • SBS and ANSI standard footprint design for broad instrument compatibility
  • Ideal for pharmaceutical applications, sample collection and storage,