VWR® Short Thread Vials ND9
Leverandør: VWR Collection
548-2644AEA 12550 NOK
548-2644A
VWR® Short Thread Vials ND9
Vialer
The vials can be used on all common autosamplers due to their technical geometry.
  • 1st hydrol. class, label and filling lines
  • Top bonded