SPE plates, 96-well, HyperSep™, Retain-AX
Leverandør: Thermo Fisher Scientific
60303-408
549-0683EA 5200 NOK
549-0683
SPE plates, 96-well, HyperSep™, Retain-AX
Mikroplater Solid fase ekstaksjonsplater
HyperSep Retain-AX is a high purity, highly porous polystyrene DVB material partially modified with quaternary functional groups to retain acidic analytes. The problems associated with column drying in traditional silica-based SPE materials are eliminated when using this product.
  • Easy recoveries for acidic analytes
  • High and consistent recoveries
  • High capacity material