n-Pentan 99% (by GC), AR®, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger

5501-25EA 2575 NOK
5501-25 5501-45
n-Pentan 99% (by GC), AR®, Macron Fine Chemicals™
n-Pentane

NB: Preserve in tight and cool conditions.

Formel: H₃C(CH₂)₃CH₃
Kokepunkt: 36,1 °C
Tetthet: 0,63
Flash punkt: –48 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009498
CAS-nummer: 109-66-0
FN: 1265
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Assay (GC) ≥99,0%
Water (KF) ≤0,02%
Titrable Acid (meq/g) ≤0,0005
Residue after Evaporation ≤0,001%
Sulfur Compounds ≤0,001%
For Laboratory, Research or Manufacturing Use


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR