Pyruvate oxidase
Leverandør: Merck Millipore Calbiochem
550701-100
550701-100EA 4250 NOK
550701-100
Pyruvate oxidase
Enzymer
Pyruvate oxidase from Aerococcus viridans catalyses the oxidation of pyruvate to acetyl phosphate, carbon dioxide, and hydrogen peroxide in the presence of phosphate.
  • Lyophilized yellow solid