96-well plate, Bond Elut™ Certify II
Leverandør: VARIAN
7541801C
551-0435EA 13700 NOK
551-0435
96-well plate, Bond Elut™ Certify II
Mikroplater Solid fase ekstaksjonsplater
Bond Elut Certify II is an anion exchange/mixed-mode SPE product suitable for extraction of acidic compounds from samples.
  • Ideal for nonpolar and anionic compounds
  • Optimised for acidic drug analysis
  • Bimodal, nonpolar and strong anion exchange
Bond Elut Certify II was developed specifically for the rapid and effective extraction of acidic drugs and metabolites from urine and other biological matrices. Certify II is a mixed-mode cartridge packed with nonpolar C8 and strong anion-exchange (SAX) sorbent.
It has been optimised for acidic drugs, such as 1More Product Information