Metalloid and non-metal mix for ICP, 10 elements, TraceCERT®, Supelco®
Leverandør: Merck
55263-100ML
55263-100MLEA 1850 NOK
55263-100ML
Metalloid and non-metal mix for ICP, 10 elements, TraceCERT®, Supelco®
Standarder Kationstandarder Multi-Element Standards
100 mg/l: B, Si, P, S, Ge, As, Se, Sn, Sb and Te in 10% nitric acid and ≤0.5% hydrofluoric acid
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034