Pliers for stainless steel tubing
Leverandør: VICI JOUR RESEARCH
JR-796
554-1105EA 935 NOK
554-1105
Pliers for stainless steel tubing
Nebbtenger
The pliers type of cutter can be used for quick cuts for non-critical applications or at hard to get places.
  • For easy cutting of ¹/₁₆" tubing off a coil