Tubing clip
Leverandør: VICI JOUR RESEARCH
JR-M9001
554-1125EA 655 NOK
554-1125
Tubing clip
Klemmer Slangeklemmer
This is a convenient small device to keep the solvent line in the solvent.
  • Attaches ¹/₁₆" or ¹/₈" tubing to beakers, flasks, or bottles
  • Stainless spring for long lifetime
Wall thickness: up to 4 mm.