Tubing cutter for polymer tubing, Upchurch Scientific®
Leverandør: UPCHURCH SCIENTIFIC
A-327
554-3024EA 1025 NOK
554-3024
Tubing cutter for polymer tubing, Upchurch Scientific®
Kuttere
Designed to cut polymer tubing ¹/₁₆" and ¹/₈" O.D.
Complete with guide holes to ensure precise cutting. The cutter is durable, reliable and easy to operate.
Leveringsinformasjon: Complete unit supplied with five replacement blades.