Connectors for capillary tubing, high pressure unions, Upchurch Scientific®
Leverandør: UPCHURCH SCIENTIFIC
U-435 P-771 P-771-01 P-772 M-572 P-720 P-720-01 U-322 U-402 U-402-01 U-322-01 U-411 U-411-01 U-412 U-412-01 U-435-01 U-437 U-438
554-3257EA 1475 NOK
554-3257 554-3185 554-3186 554-3187 554-3087 554-3167 554-3168 554-3243 554-3245 554-3246 554-3244 554-3248 554-3249 554-3250 554-3251 554-3258 UPCHU-437 UPCHU-438
Connectors for capillary tubing, high pressure unions, Upchurch Scientific®
Fittings og kontakter
Stainless steel and PEEK polymer.
Frequently there is a need in the high pressure region of a system to bring two pieces of tubing together in a reliable connection. To meet that need, we engineered a number of high pressure unions in a variety of configurations.