Injection port liners for Agilent Technologies instruments
Leverandør: Thermo Fisher Scientific
453A2342 453A2002 453A2265 453A2275 453A2282 453A2292 453A1355 453A2375 453A1925 453A1252 453A1255 453A1262 453A1285 453A1295 453A1305 453A1312 453A1315 453A1335 453A1345
HYPE453A2342EA 8025 NOK
HYPE453A2342 HYPE453A2002 555-0133 HYPE453A2275 HYPE453A2282 HYPE453A2292 555-0137 HYPE453A2375 HYPE453A1925 HYPE453A1252 555-0134 HYPE453A1262 HYPE453A1285 HYPE453A1295 HYPE453A1305 HYPE453A1312 555-0135 555-0132 555-0136
Injection port liners for Agilent Technologies instruments
Kromatografiutstyr Kromatografiledninger
Optimise the performance of Agilent™ GC systems with these specifically designed injection port liners for Agilent instruments.
  • Highly deactivated
  • Produced to exacting tolerances
  • Highly deactivated borosilicate glass