Capillary column end caps
Leverandør: Thermo Fisher Scientific
260EC111
555-9468EA 1025 NOK
555-9468
Capillary column end caps
Fittings og kontakter
These capillary column end caps are universal and fit any capillary column.
  • Reusable
  • Eliminate column contamination caused by leaving a column unsealed or sealed with a septum
  • Colour coded to ensure that the column is reinstalled the same way it came out