Organic reference standard, Spermine, analytical standard, Supelco®
Leverandør: Merck
55513-100MG
55513-100MGEA 580 NOK
55513-100MG
Organic reference standard, Spermine, analytical standard, Supelco®
Standarder Organiske standarder
Mixed NMDA glutamate receptor agonist/antagonist at the polyamine site. Neuroprotective effects have been observed at high concentrations (1 mM), while neurotoxicity is observed at lower concentrations. It enhances agonist effectiveness at the strychnine-insensitive glycine site. Plays a role in cellular proliferation and differentiation, inhibits neuronal nitric oxide synthase (nNOS).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4b3b5e601e7446c7c6bf2e3b80d932f5cb46bf0&vid=edcc217ca1188bf17d6ccd75df17f4a78093fcd4&prid=web&clid=SAPDAM