Saltsyre 3.571 mol/l (N/0.28), AVS TITRINORM volumetric solution

Leverandør: VWR Chemicals
Warning

5610.1000EA 400 NOK
5610.1000 5610.9010
Saltsyre 3.571 mol/l (N/0.28), AVS TITRINORM volumetric solution
Saltsyre
Formel: HCl
Tetthet: 1,06 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR