Sodium thiophosphate hydrate

Leverandør: Thermo Fisher Scientific
Warning

56146.06 56146.14
56146.06EA 1250 NOK
56146.06 56146.14
Sodium thiophosphate hydrate
Sodium thiophosphate hydrate

CAS No.: 10489-48-2 Molecular Formula: Na3PO3S.xH2O Formula Weight: 179.93(anhy) Physical Form: Crystalline aggregates Appearance: White to green Melting Point: 53-58° MDL No.: MFCD00041513

Formel: H₂Na₃O₄PS
Smeltepunkt: 53…58 °C
Tetthet: 1,67 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00041513
CAS-nummer: 10489-48-2
EINECS: 231-867-5
Merck Indeks: 14,8693

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR