Dodecamolybdofosforsyre hydrat ACS

Leverandør: Thermo Fisher Scientific
Danger

56166.22 56166.06 56166.14 56166.36
56166.22EA 2200 NOK
56166.22 56166.06 56166.14 56166.36
Dodecamolybdofosforsyre hydrat ACS
Dodecamolybdofosforsyre hydrat

CAS No.: 51429-74-4 Molecular Formula: H3PO4.12MoO3.xH2O Formula Weight: 1825.25(anhy) Physical Form: Granular Density: 3.15 Sensitivity: Moisture Sensitive Appearance: Yellow Melting Point: 78-90° Merck Reference: 14,7342 UN No.: 3085 MDL No.: MFCD00149913

Formel: H₃[P(Mo₃O₁₀)₄]·nH₂O
Smeltepunkt: 78…90 °C
Tetthet: 3,15
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149913
CAS-nummer: 51429-74-4
EINECS: 234-713-5
FN: 3085
ADR: 5.1,III
Merck Indeks: 14,7342

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR