Maursyre ( metansyre) ≥97.5% eluent additive, for LC-MS, Fluka™

Leverandør: Honeywell Chemicals
Danger

Synonymer: Aminic Acid, Methanoic acid

56302-10X1ML 56302-1L-GL 56302-50ML-GL
56302-10X1MLEA 1350 NOK
56302-10X1ML 56302-1L-GL 56302-50ML-GL
Maursyre ( metansyre) ≥97.5% eluent additive, for LC-MS, Fluka™
Maursyre ( metansyre)
Formel: HCO₂H
MW: 46.03 g/mol
Kokepunkt: 100.4 °C (1.013 hPa)
Smeltepunkt: 4 °C
Tetthet: 1.22 g/cm³ (20 °C)
Flash punkt: 49.5 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003297
CAS-nummer: 64-18-6
EINECS: 200-579-1
FN: 1779
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay Min. 97.5 %
Refractive index (n 20/D) 1.370 - 1.372
Non-volatile matter Max. 0.002 %
Silver (Ag) Max. 0.02 ppm
Aluminium (Al) Max. 0.05 ppm
Barium (Ba) Max. 0.05 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.2 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.05 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.2 ppm
Potassium (K) Max. 0.1 ppm
Lithium (Li) Max. 0.02 ppm
Magnesium (Mg) Max. 0.5 ppm
Manganese (Mn) Max. 0.05 ppm
Molybdenum (Mo) Max. 0.02 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.05 ppm
Lead (Pb) Max. 0.02 ppm
Strontium (Sr) Max. 0.02 ppm
Thallium (Tl) Max. 0.05 ppm
Zinc (Zn) Max. 0.05 ppm
Ammonium (NH₄) Max. 10 ppm
Chloride (Cl) Max. 5 ppm
Sulfate (SO₄) Max. 5 ppm
Sulfite (SO₃) Max. 10 ppm
Suitability for the LC-MS complying
Appearance of the solution complying
APHA Max. 15
Vapor Density 1.6 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR