Air compressor, Solaris
Leverandør: PEAK SCIENTIFIC
65-0003
564-0278EA 63900 NOK
564-0278
Air compressor, Solaris
Gassgeneratorer Trykkluftstasjoner
This air compressor generator has been designed to support the Solaris nitrogen generator for ELSD in laboratories without an external supply of high quality air.
  • Stackable with Solaris nitrogen generator for ELSD and Compact Mass Specs
  • Designed specifically for ELSD and Compact Mass Specs
Solaris has been developed to provide an on-site gas supply of high purity nitrogen for ELSD for laboratories using HPLC or alternatively to supply Compact Mass Spectrometer instruments.
Min./max. operating temperature: 5 to 35 °C
Max. pressure: 8,27 bar