Nitrogen and dry air gas generator, Genius 3045
Leverandør: PEAK SCIENTIFIC
10-3045
564-0280EA 0 NOK
564-0280
Nitrogen and dry air gas generator, Genius 3045
Gassgeneratorer Nitrogengeneratorer
Designed to the specifications of the Bruker EVOQ triple Quadrupole's gas requirements delivering dry air at up to 50 L/min and nitrogen at up to 32 L/min.
  • Independent nitrogen and dry air gas supplies can be tailored for its application
  • Higher flow 30 series generator designed for hungrier LC-MS applications
  • Compressor based solution, no need for an external air supply
  • Highly economical source of nitrogen gas with low lifetime running costs
  • Gas is supplied on demand so generator works to your schedule
  • Service indication to allow planning of preventative