Nitrogen generator, Precision Nitrogen Headspace 250
Leverandør: PEAK SCIENTIFIC
65-3000
564-0298EA 70100 NOK
564-0298
Nitrogen generator, Precision Nitrogen Headspace 250
Gassgeneratorer Nitrogengeneratorer
The Precision Nitrogen Headspace 250 has been conceived and custom-engineered to meet the gas requirements for headspace sampler instruments requiring higher gas pressure.
  • Ultra High Purity nitrogen for headspace sampler instruments
  • Can supply make-up gas
  • Single unit can stack with the Precision Hydrogen, Nitrogen and Zero Air generators
  • Service and repair status indicators
  • 1 year comprehensive on-site warranty