1,5-Diaminonaftalen 99,0 HPLC_ASSAY_METHOD MALDI-MS Matrix, Supelco®

Leverandør: Merck
Warning

Synonymer: 1,5-naphthylenediamine, 1,5-Naphthalenediamine

56451-250MG
56451-250MGEA 1180 NOK
56451-250MG
1,5-Diaminonaftalen 99,0 HPLC_ASSAY_METHOD MALDI-MS Matrix, Supelco®
1,5-Diaminonaftalen

1,5-Diaminonaphthalene is used as an effective matrix in the analysis of IgG protein using top-down and middle-down mass spectrometry with multiple fragmentation techniques such as electron transfer dissociation (ETD) and matrix-assisted laser desorption ionization in-source decay (MALDI-ISD) method.


1,5-Diaminonaphthalene is used in the production of 1,5-naphthalene diisocyanate and polyurethane elastomers.


Forpakning: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₁₀H₁₀N₂
MW: 158.2 g/mol
Kokepunkt: 381
Smeltepunkt: 188
MDL nummer: MFCD00004029
CAS-nummer: 2243-62-1
FN: 3077
ADR: 9 III

Bestill nå

Spesifikasjon Testresultater

Appearance (Color) White to Beige and Faint Brown to Brown and Faint Purple to Light Purple
Appearance (Form) Powder to Crystals
Purity (HPLC area %) ≥ 99.0 %
Identity MALDI-MS suitable as Maldi Matrix Material
Carbon Content 75.92 % (Theory)
Hydrogen Content 6.37 % (Theory)
Nitrogen Content 17.71 % (Theory)
Proton NMR Spectrum Conforms to structure
Metal trace Analysis (ICP) Corresponds to requirements
BARIUM (ICP) ≤ 5.0 mg/kg
CALCIUM (ICP) ≤ 5.0 mg/kg
CADMIUM (ICP) ≤ 5.0 mg/kg
COBALT (ICP) ≤ 5.0 mg/kg
CHROMIUM (ICP) ≤ 5.0 mg/kg
COPPER (ICP) ≤ 5.0 mg/kg
POTASSIUM (ICP) ≤ 10 mg/kg
MAGNESIUM (ICP) ≤ 5.0 mg/kg
MANGANESE (ICP) ≤ 5.0 mg/kg
SODIUM (ICP) ≤ 10 mg/kg
NICKEL (ICP) ≤ 5.0 mg/kg
LEAD (ICP) ≤ 5.0 mg/kg
ZINC (ICP) ≤ 5.0 mg/kg
RECOMMENDED RETEST PERIOD 36 MONTHS

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR