Sølvnitrat standard solution (1 mL = 1 mg Chloride), BAKER ANALYZED®, J.T.Baker®

Leverandør: Avantor
Warning

Synonymer: Lunar caustic, Silver mononitrate, Nitric acid silver salt

5652-03EA 0 NOK
5652-03
Sølvnitrat standard solution (1 mL = 1 mg Chloride), BAKER ANALYZED®, J.T.Baker®
Sølvnitrat

Standardized at 25°C against chemicals whose certification is traceable to NIST.

Formel: AgNO₃
MW: 169.87 g/mol
Kokepunkt: 444
Smeltepunkt: 210
MDL nummer: MFCD00003414
CAS-nummer: 7761-88-8
Merck Indeks: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use
Titer, mg/mL, For Information Only 3.0 - 3.5
pH at 25°C 3.0 - 3.5
Sulfate (SO₄) ≤ 1 ppm
Each lot of this product is standardized at 25°C against ULTREX® SodiumChloride, which is traceable to NIST Standard Reference Materials, withpotassium chromate indication.
SRM NoÖ
The titer found falls within the range 0.995-1.005 mg Cl/mL.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR