Natriumhydroksid 6 N vandig løsning, BAKER ANALYZED® volumetrisk løsning, J.T.Baker®

Leverandør: Avantor
Danger

5672-02EA 1025 NOK
5672-02
Natriumhydroksid 6 N vandig løsning, BAKER ANALYZED® volumetrisk løsning, J.T.Baker®
Natriumhydroksid

J.T.Baker® volumetric and analytical solutions. These solutions are suitable for use in ACS, USP and NF compendial methods and general laboratory applications.

Formel: NaOH
Kokepunkt: 109 °C (1013 hPa)
Tetthet: ≤1,11 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
FN: 1824
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use 5.98 - 6.02
Protect from air to avoid absorption of carbon dioxide. Max. 5 ppm
Normality 5.98 - 6.02
Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR