Spanner, open end, SGE
Leverandør: Trajan Scientific and Medical
18500002
568-0686EA 550 NOK
568-0686
Spanner, open end, SGE
Tang
Spanner sizes and uses: 3/16" for use with mini unions, 1/4" for use with 1/16" nuts, 5/16" for use with micro control valves.