Mopping systems for cleanrooms
Leverandør: CONTEC
2787
568-0903EA 25625 NOK
568-0903
Mopping systems for cleanrooms
Mopper
This mop head saturation system allows mop heads to be pre-saturated prior to being passed into a controlled environment. This eliminates the need for any water to be passed into the cleanroom.
  • 100% 2 ply polyester covered mop heads for low levels of particles and fibres
  • Stainless steel trolleys with PP castors
  • Hands-free mop application eliminates the need for a heavy or expensive wringer system
  • Pre-saturated mop heads means no water needs to be taken into the cleanroom
  • Compatible with a wide range of disinfectants and detergents
  • Trolleys are suitable for