Copper (II) ethylenediamine 1 mol/l, VOLUSOL®

Leverandør: VWR Chemicals
Danger

Synonymer: Kobber(II)etylendiaminiløsning

5761.5000EA 1600 NOK
5761.5000
Copper (II) ethylenediamine 1 mol/l, VOLUSOL®
Copper (II) ethylenediamine
Formel: Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
MW: 217.76 g/mol
MDL nummer: MFCD00274628
CAS-nummer: 14552-35-3
EINECS: 238-597-7
FN: 1761
ADR: 8 (6.1) II

Bestill nå

Spesifikasjon Testresultater

Density (20/4) 1.096 - 1.102
Copper 0.98 - 1.02 mol/l
Mol proportion (Ethylenediamine/Cu) 1.90 - 2.10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR