orto-Borsyre 40 g/l in aqueous solution

Leverandør: VWR Chemicals

Synonymer: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

5792.5000EA 1275 NOK
5792.5000
orto-Borsyre 40 g/l in aqueous solution
orto-Borsyre
Formel: B(OH)₃
Tetthet: 1,0174 g/cm³
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011337
CAS-nummer: 10043-35-3
EINECS: 233-139-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay 3.9 - 4.1 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR