VWR®, Safety Waste Caps
Leverandør: VWR Collection
590-0043EA 1700 NOK
590-0043 590-0044 590-0045 590-0046
VWR®, Safety Waste Caps
Lokk Lokk, sikkerhetsavfall
Collect your solvent waste safely without being exposed to solvent vapours or hazardous gases. High quality materials (PTFE, PP) ensure maximum safety for your laboratory staff and environment.
  • Recommended for HPLC systems
  • Chemically resistant against aggressive organic solvents
  • Tubing connector for e.g. leak connection of diverse HPLC instruments (only for safety waste caps with tube connections)
Bestillingsinformasjon: Exhaust air filter needs to be ordered separately.