Exhaust filters, b.safe
Leverandør: Bohlender
M506-02 M506-01 M506-03
590-0411EA 3000 NOK
590-0411 590-0176 590-0178
Exhaust filters, b.safe
Filter Ventilasjonsfilter
Filter body made of PP with connector (thread GL 14) for b.safe waste caps. Lid with aeration openings and service life indicator with additional label field to monitor service life. Filling made of activated carbon for adsorption of solvents.
  • Activation of service life through touch of the button and/or individual marking on the back side
  • Splash-proof ventilation openings
  • Activated carbon in the inside guarantee adsorption of harmful vapours
  • PP for high resistance and fire class rating V-0
  • Plenty of space for fittings and hose connectors