Distributor for capillaries, b.safe
Leverandør: Bohlender
M511-01
590-0195EA 1100 NOK
590-0195
Distributor for capillaries, b.safe
Fittings og kontakter
Distributor for connection of capillary tubing. With PFA Fittings for capillaries with Ext.Ø 3,2 mm (thread UNF ¹/₄").
  • Excellent chemical resistance
  • Available for a wide range of hose diameters
  • The conical shape gets the connection tight and leak-free
Adaptor to enlarge the number of capillary connectors on b.safe Caps.