Capillary connection, b.safe
Leverandør: Bohlender
M512-01
590-0196EA 615 NOK
590-0196
Capillary connection, b.safe
Fittings og kontakter
Straight fitting made of PTFE, capillary connection on both sides (UNF ¹/₄" thread), with 2 PFA fittings each for tubing Ext.Ø 1,6/2,2/3,2 mm.
  • Excellent chemical resistance
  • Available for a wide range of hose diameters
  • The conical shape gets the connection tight and leak-free
Adaptor for connecting capillaries of the same size.