Tubing holder, b.safe
Leverandør: Bohlender
M562-01
590-0220EA 365 NOK
590-0220
Tubing holder, b.safe
Klemmer Slangeklemmer
For fixation of capillaries and tubing to avoid disarranging or bending of tubes. Self-adhesive bottom side.
  • Avoid disarranging or bending of tubes
  • Lead and fix b.safe tubes
  • Made of ABS
A b.safe tubing holder can arrange at once up to 8× Ext.Ø 1,6 mm / 3× Ext.Ø 3,2 mm or 1× Ext.Ø 5 mm.
To lead and fix tubing and capillaries without being disarranged or bent.