Canister, S50, b.safe
Leverandør: Bohlender
R210-05
590-0290EA 1200 NOK
590-0290
Canister, S50, b.safe
Sikkerhetskanner
Space-saving canister with S50 thread (KS50) for the collection of liquid waste.
  • Including screw cap
  • Canister for space-saving
  • b.safe accessories such as collecting trays, spouts and grounding cables, for dissipative canisters available
b.safe offers also suitable containers like flasks and canisters in addition to the wide range of caps for the extraction of solvents and waste caps or funnels for the collection of liquid waste.