HPLC All-In-One box, b.safe
Leverandør: Bohlender
R9900-10
590-0412EA 9475 NOK
590-0412
HPLC All-In-One box, b.safe
Kromatografiutstyr Kromatografireservedeler
Complete set for equipping one HPLC system.
Containing b.safe Caps GL 45 for four laboratory flasks for solvent extraction as well as a b.safe waste cap with exhaust filter and a canister for safe collection of solvent waste. The scope of delivery of the b.safe caps includes PFA fittings for capillary Ext.Ø. 3,2 mm, blind fitting in PFA to close unused connections and Air Valves with service life indicator. For the collection of waste, the set includes a PE-canister S55 with 5-litre capacity, a suitable waste cap S55 and one exhaust filter L. The scope of delivery of the waste capMore Product Information